4-Isocyanato-5-methyl-3-phenylisoxazole

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-Isocyanato-5-methyl-1,2-oxazol-3-yl)benzene, 4-Isocyanato-5-methyl-3-phenyl-1,2-oxazole

OR22981-1G
APOSOR22981-1GEA 328 EUR
APOSOR22981-1G
4-Isocyanato-5-methyl-3-phenylisoxazole
4-Isocyanato-5-methyl-3-phenylisoxazole

When melted, may contain small amount of sediment

Formel: C₁₁H₈N₂O₂
Molecular Weight: 200.193 g/mol
Siedepunkt: 108…110 °C (0.8 mm Hg)
Schmelzpunkt: 39…40 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02677698
CAS-Nummer: 268748-84-1
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR