4-(Brommethyl)-5-methyl-3-phenylisoxazol 97%

Lieferant: Apollo Scientific

OR22983-250MG
APOSOR22983-250MGEA 132 EUR
APOSOR22983-250MG
4-(Brommethyl)-5-methyl-3-phenylisoxazol 97%
4-(Bromomethyl)-5-methyl-3-phenylisoxazole
Formel: C₁₁H₁₀BrNO
Schmelzpunkt: 61…63 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02677680
CAS-Nummer: 180597-83-5
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR