3,5-Dimethyl-1,2-oxazol-4-carbaldehyd

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,5-Dimethylisoxazole-4-carboxaldehyde, 3,5-Dimethylisoxazol-4-carboxaldehyde

OR22987-250MG
APOSOR22987-250MGEA 21.9 EUR
APOSOR22987-250MG
3,5-Dimethyl-1,2-oxazol-4-carbaldehyd
3,5-Dimethyl-4-isoxazolecarbaldehyde
Formel: C6H7NO2
Molecular Weight: 125,13 g/mol
Siedepunkt: 54…56 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02681977
CAS-Nummer: 54593-26-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR