1,4-Diazepan-5-one hydrochloride 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Homopiperazin-5-one hydrochloride

OR2299-1G
APOSOR2299-1GEA 23.2 EUR
APOSOR2299-1G
1,4-Diazepan-5-one hydrochloride 98%
1,4-Diazepan-5-one hydrochloride
Formel: C₅H₁₁ClN₂O
Molecular Weight: 150,6066 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09864983
CAS-Nummer: 208245-76-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR