2,3-Dihydro-1-benzofuran-5-sulfonylchlorid
Lieferant: Apollo Scientific
|
Gefahr
|
OR22992-1G
OR22992-10G
OR22992-500MG
APOSOR22992-1GEA
102
EUR
APOSOR22992-1G
APOSOR22992-10G
APOSOR22992-500MG
2,3-Dihydro-1-benzofuran-5-sulfonylchlorid
2,3-Dihydro-1-benzofuran-5-sulphonoylchloride
Formel:
C₈H₇ClO₃S Schmelzpunkt: 74…85 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02677703 CAS-Nummer: 115010-11-2 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...