5-Brom-2,3-dihydro-1-benzofuran

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromocoumaran

OR22995-1G
APOSOR22995-1GEA 21.9 EUR
APOSOR22995-1G
5-Brom-2,3-dihydro-1-benzofuran
5-Brom-2,3-dihydro-1-benzofuran
Formel: C₈H₇BrO
Molecular Weight: 199,05 g/mol
Schmelzpunkt: 49…50 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02677716
CAS-Nummer: 66826-78-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR