3,5-Dimethyl-1H-pyrazole-4-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3,5-Dimethylpyrazole-4-carboxylic Acid
OR23000-1G
APOSOR23000-1GEA
24.8
EUR
APOSOR23000-1G
3,5-Dimethyl-1H-pyrazole-4-carboxylic acid
3,5-Dimethyl-1H-pyrazole-4-carboxylic acid
Formel:
C6H8N2O2 Schmelzpunkt: 273…275 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00159642 CAS-Nummer: 113808-86-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...