3,5-Dimethyl-4-iod-1H-pyrazol 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3,5-Dimethyl-4-iodopyrazole

OR23002-100G
APOSOR23002-100GEA 68.3 EUR
APOSOR23002-100G
3,5-Dimethyl-4-iod-1H-pyrazol 97%
3,5-Dimethyl-4-iodo-1H-pyrazole
Formel: C₅H₇IN₂
Molecular Weight: 222.03 g/mol
Schmelzpunkt: 137…140 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00040247
CAS-Nummer: 2033-45-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR