2,3-Dihydrobenzo[1,4]dioxine-6-sulphonyl chloride

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2,3-Dihydrobenzo[b][1,4]dioxine-6-sulphonyl chloride

OR23005-5G
APOSOR23005-5GEA 0 EUR
APOSOR23005-5G
2,3-Dihydrobenzo[1,4]dioxine-6-sulphonyl chloride
2,3-Dihydrobenzo[1,4]dioxine-6-sulphonyl chloride
Formel: C₈H₇ClO₄S
Molecular Weight: 234.66 g/mol
Schmelzpunkt: 65…71 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02677681
CAS-Nummer: 63758-12-3
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR