4-Brom-1,3-benzodioxol

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromobenzo[d][1,3]dioxole

OR23021-250MG
APOSOR23021-250MGEA 79.9 EUR
APOSOR23021-250MG
4-Brom-1,3-benzodioxol
4-Brom-1,3-benzodioxol
Formel: C₇H₅BrO₂
Molecular Weight: 201,0174 g/mol
Siedepunkt: 82…83 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02681890
CAS-Nummer: 6698-13-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR