2-Bromfuran
Lieferant: Apollo Scientific
OR23025-1G
APOSOR23025-1GEA
65.4
EUR
APOSOR23025-1G
2-Bromfuran
2-Bromofuran
Formel:
C₄H₃BrO |
MDL:
MFCD00216609 CAS-Nummer: 584-12-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...