3-Furylmethanol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Furyl Alcohol, 3-(Hydroxymethyl)furan
OR23029-25G
APOSOR23029-25GEA
213
EUR
APOSOR23029-25G
3-Furylmethanol
3-Furylmethanol
Formel:
C₅H₆O₂ Molecular Weight: 98.10 g/mol Siedepunkt: 79…80 °C/17mmHg Flammpunkt: 38 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005352 CAS-Nummer: 4412-91-3 UN: 1987 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...