2-Bromo-1-(5-pyridin-2-ylthiophen-2-yl)ethanone 90%

Lieferant: Apollo Scientific
Achtung

OR23034-1G
APOSOR23034-1GEA 0 EUR
APOSOR23034-1G
2-Bromo-1-(5-pyridin-2-ylthiophen-2-yl)ethanone 90%
2-Bromo-1-(5-pyridin-2-ylthiophen-2-yl)ethanone
Formel: C₁₁H₈BrNOS
Molecular Weight: 282,17 g/mol
Schmelzpunkt: 123…126 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02677697
CAS-Nummer: 306935-06-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR