2-(Hydroxymethyl)-5-(pyridin-2-yl)thiophene

Lieferant: Apollo Scientific
Achtung

Synonyme: [5-(Pyridin-2-yl)thien-2-yl]methanol

OR23035-100MG
APOSOR23035-100MGEA 197 EUR
APOSOR23035-100MG
2-(Hydroxymethyl)-5-(pyridin-2-yl)thiophene
2-(Hydroxymethyl)-5-(pyridin-2-yl)thiophene
Formel: C₁₀H₉NOS
Molecular Weight: 191,25 g/mol
Schmelzpunkt: 85…87 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02677743
CAS-Nummer: 197899-76-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR