1-(Phenylsulphonyl)-1H-indole-3-carboxaldehyde 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(Phenylsulphonyl)-1H-indole-3-carbaldehyde, 3-Formyl-1-(phenylsulphonyl)-1H-indole

OR23044-1G
APOSOR23044-1GEA 101 EUR
APOSOR23044-1G
1-(Phenylsulphonyl)-1H-indole-3-carboxaldehyde 97%
1-(Phenylsulphonyl)-1H-indole-3-carboxaldehyde
Formel: C₁₅H₁₁NO₃S
Molecular Weight: 285,318 g/mol
Schmelzpunkt: 157…161 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02681985
CAS-Nummer: 80360-20-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR