1-(Phenylsulphonyl)-1H-indole-2-carbonyl chloride

Lieferant: Apollo Scientific
Gefahr

OR23049-500MG
APOSOR23049-500MGEA 109 EUR
APOSOR23049-500MG
1-(Phenylsulphonyl)-1H-indole-2-carbonyl chloride
1-(Phenylsulphonyl)-1H-indole-2-carbonyl chloride
Formel: C₁₅H₁₀ClNO₃S
Molecular Weight: 319,763 g/mol
Schmelzpunkt: 127 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03086090
CAS-Nummer: 342405-28-1
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR