8-(Brommethyl)chinolin
Lieferant: Apollo Scientific
|
Gefahr
|
OR23071-1G
APOSOR23071-1GEA
24.8
EUR
APOSOR23071-1G
8-(Brommethyl)chinolin
8-(Bromomethyl)quinoline
Formel:
C₁₀H₈BrN Schmelzpunkt: 81…82 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00047621 CAS-Nummer: 7496-46-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...