2-Methyl-4-thiazolcarbonsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Methylthiazole-4-carboxylic acid

OR23080-1G
APOSOR23080-1GEA 21.9 EUR
APOSOR23080-1G
2-Methyl-4-thiazolcarbonsäure
2-Methyl-4-thiazolecarboxylic acid
Formel: C₅H₅NO₂S
Molecular Weight: 143,1637 g/mol
Schmelzpunkt: 145…150 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03407332
CAS-Nummer: 35272-15-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR