1,3-Benzothiazole-2-carbaldehyde 98%

Lieferant: Apollo Scientific
Achtung

OR23088-5G
APOSOR23088-5GEA 92.8 EUR
APOSOR23088-5G
1,3-Benzothiazole-2-carbaldehyde 98%
1,3-Benzothiazole-2-carbaldehyde
Formel: C₈H₅NOS
Molecular Weight: 163.2 g/mol
Schmelzpunkt: 72…76 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00526215
CAS-Nummer: 6639-57-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR