Benzothiazol-2-ylmethanol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(Hydroxymethyl)benzothiazole, 2-Benzothiazolemethanol, 2-Hydroxymethylbenzothiazole
OR23091-250MG
APOSOR23091-250MGEA
21.9
EUR
APOSOR23091-250MG
Benzothiazol-2-ylmethanol
Benzothiazol-2-ylmethanol
Formel:
C₈H₇NOS Schmelzpunkt: 95…100 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00226293 CAS-Nummer: 37859-42-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...