1-[(Benzyloxy)carbonyl]-4-piperidincarbonsäure 97%

Lieferant: Apollo Scientific
Achtung

OR23101-1G
APOSOR23101-1GEA 21.9 EUR
APOSOR23101-1G
1-[(Benzyloxy)carbonyl]-4-piperidincarbonsäure 97%
N-Cbz-4-piperidinecarboxylic acid
Formel: C₁₄H₁₇NO₄
Schmelzpunkt: 76…81 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01568759
CAS-Nummer: 10314-98-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR