Piperidine-4-carbonyl chloride, N-CBZ protected

Lieferant: Apollo Scientific
Gefahr

OR23102-1G OR23102-250MG OR23102-500MG
APOSOR23102-1GEA 184 EUR
APOSOR23102-1G APOSOR23102-250MG APOSOR23102-500MG
Piperidine-4-carbonyl chloride, N-CBZ protected
Piperidine-4-carbonyl chloride, N-CBZ protected
Formel: C₁₄H₁₆ClNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD02677707
CAS-Nummer: 10314-99-5
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR