Benzyl-4-(hydroxymethyl)piperidine-1-carboxylate 97%

Lieferant: Apollo Scientific

OR23106-25G
APOSOR23106-25GEA 0 EUR
APOSOR23106-25G
Benzyl-4-(hydroxymethyl)piperidine-1-carboxylate 97%
Benzyl-4-(hydroxymethyl)piperidine-1-carboxylate
Formel: C₁₄H₁₉NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD02094490
CAS-Nummer: 122860-33-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR