Pyrrol-2-carbonsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR23111-5G
APOSOR23111-5GEA
60.9
EUR
APOSOR23111-5G
Pyrrol-2-carbonsäure 97%
Pyrrol-2-carbonsäure
Formel:
C₅H₅NO₂ Molecular Weight: 111,10 g/mol Schmelzpunkt: 204…209 °C (Subl.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005219 CAS-Nummer: 634-97-9 EINECS: 211-221-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...