1H-Imidazole-4-thiocarboxamide 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Carbamothioyl-1H-imidazole, 1H-Imidazole-4-carbothioic acid amide

OR23114-250MG
APOSOR23114-250MGEA 52.2 EUR
APOSOR23114-250MG
1H-Imidazole-4-thiocarboxamide 97%
1H-Imidazole-4-thiocarboxamide
Formel: C₄H₅N₃S
Molecular Weight: 127,168 g/mol
Schmelzpunkt: 204…207 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00233357
CAS-Nummer: 95962-95-1
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR