2,1,3-Benzothiadiazol-4-carbaldehyd

Lieferant: Apollo Scientific
Achtung

Synonyme: Benzo[c][1,2,5]thiadiazole-4-carbaldehyde

OR23119-1G
APOSOR23119-1GEA 85.7 EUR
APOSOR23119-1G
2,1,3-Benzothiadiazol-4-carbaldehyd
2,1,3-Benzothiadiazol-4-carbaldehyd
Formel: C₇H₄N₂OS
Molecular Weight: 164.1845 g/mol
Schmelzpunkt: 101…102 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02681902
CAS-Nummer: 5170-68-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR