2,1,3-Benzothiadiazol-4-ylisocyanat 95%

Lieferant: Apollo Scientific
Gefahr

OR23120-500MG
APOSOR23120-500MGEA 0 EUR
APOSOR23120-500MG
2,1,3-Benzothiadiazol-4-ylisocyanat 95%
2,1,3-Benzothiadiazol-4-ylisocyanat

When melted, may contain small amount of sediment.

Formel: C₇H₃N₃OS
Schmelzpunkt: 81…85 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02681903
CAS-Nummer: 342411-14-7
UN: 2206
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR