2,1,3-Benzothiadiazol-5-carbonylchlorid

Lieferant: Apollo Scientific
Gefahr

OR23125-1G OR23125-250MG
APOSOR23125-1GEA 0 EUR
APOSOR23125-1G APOSOR23125-250MG
2,1,3-Benzothiadiazol-5-carbonylchlorid
2,1,3-Benzothiadiazol-5-carbonylchlorid
Formel: C₇H₃ClN₂OS
Lagertemperatur: Raumtemperatur
MDL: MFCD02681992
CAS-Nummer: 321309-31-3
UN: 1759
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR