4-(Brommethyl)-5-phenyloxazol 95%
Lieferant: Apollo Scientific
|
Gefahr
|
OR23139-100MG
APOSOR23139-100MGEA
171
EUR
APOSOR23139-100MG
4-(Brommethyl)-5-phenyloxazol 95%
4-(Bromomethyl)-5-phenyloxazole
Formel:
C₁₀H₈BrNO Schmelzpunkt: 50…52 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03407316 CAS-Nummer: 368869-94-7 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...