5-Methyl-3-isoxazolcarbonsäure 97%

Lieferant: Apollo Scientific
Achtung

OR23141-25G
APOSOR23141-25GEA 55.1 EUR
APOSOR23141-25G
5-Methyl-3-isoxazolcarbonsäure 97%
5-Methylisoxazole-3-carboxylic acid
Formel: C₅H₅NO₃
Molecular Weight: 127,1 g/mol
Schmelzpunkt: 168…173 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01318162
CAS-Nummer: 3405-77-4
EINECS: 222-289-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR