(5-Methylisoxazol-3-yl)methylamine hydrochloride
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3-(Aminomethyl)-5-methylisoxazole
OR23144-1G
APOSOR23144-1GEA
0
EUR
APOSOR23144-1G
(5-Methylisoxazol-3-yl)methylamine hydrochloride
(5-Methylisoxazol-3-yl)methylamine hydrochloride
Formel:
C₅H₉ClN₂O Molecular Weight: 148,59 g/mol Siedepunkt: 75…76 °C/0,4mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD02681995 CAS-Nummer: 154016-48-5 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...