5-Phenyl-2-thiophenboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: (5-Phenylthiophen-2-yl)boronic acid

OR23152-1G
APOSOR23152-1GEA 37.7 EUR
APOSOR23152-1G
5-Phenyl-2-thiophenboronsäure
5-Phenyl-2-thiopheneboronic acid
Formel: C₁₀H₉BO₂S
Molecular Weight: 204,0500 g/mol
Schmelzpunkt: 140 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01114643
CAS-Nummer: 306934-95-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR