2-(2-Thienyl)-4-thiazolcarbonsäure

Lieferant: Apollo Scientific

Synonyme: 2-(2-Thienyl)-1,3-thiazole-4-carboxylicacid

OR23153-1G
APOSOR23153-1GEA 248 EUR
APOSOR23153-1G
2-(2-Thienyl)-4-thiazolcarbonsäure
2-(2-Thienyl)-4-thiazolecarboxylic acid
Formel: C₈H₅NO₂S₂
Molecular Weight: 211,25 g/mol
Schmelzpunkt: 156…160 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01936014
CAS-Nummer: 24044-07-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR