5-(Thiophen-2-yl)nicotinic acid 97%

Lieferant: Apollo Scientific
Achtung

OR23158-1G OR23158-250MG
APOSOR23158-1GEA 219 EUR
APOSOR23158-1G APOSOR23158-250MG
5-(Thiophen-2-yl)nicotinic acid 97%
5-(Thiophen-2-yl)nicotinic acid
Formel: C₁₀H₇NO₂S
Schmelzpunkt: 260…261 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02682027
CAS-Nummer: 306934-96-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR