5-(Thiophen-2-yl)nicotinic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR23158-1G
OR23158-250MG
APOSOR23158-1GEA
219
EUR
APOSOR23158-1G
APOSOR23158-250MG
5-(Thiophen-2-yl)nicotinic acid 97%
5-(Thiophen-2-yl)nicotinic acid
Formel:
C₁₀H₇NO₂S Schmelzpunkt: 260…261 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02682027 CAS-Nummer: 306934-96-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...