4-(Thiophen-2-yl)benzoic acid

Lieferant: Apollo Scientific

OR23163-5G
APOSOR23163-5GEA 143 EUR
APOSOR23163-5G
4-(Thiophen-2-yl)benzoic acid
4-(Thiophen-2-yl)benzoic acid
Formel: C₁₁H₈O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD00690742
CAS-Nummer: 29886-62-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR