2-Benzo[b]thiophensulfonylchlorid
Lieferant: Apollo Scientific
|
Gefahr
|
OR23168-250MG
APOSOR23168-250MGEA
43.5
EUR
APOSOR23168-250MG
2-Benzo[b]thiophensulfonylchlorid
2-Benzo[b]thiophensulfonylchlorid
Formel:
C₈H₅ClO₂S₂ Schmelzpunkt: 74…76 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03659701 CAS-Nummer: 90001-64-2 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...