Benzo[b]thiophen-2-methanol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(Hydroxymethyl)benzo[b]thiophene, Benzo[b]thiophen-2-ylmethanol
OR23169-500MG
APOSOR23169-500MGEA
21.9
EUR
APOSOR23169-500MG
Benzo[b]thiophen-2-methanol
Benzo[b]thiophen-2-methanol
Formel:
C₉H₈OS Molecular Weight: 164,22 g/mol Schmelzpunkt: 99…103 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02682004 CAS-Nummer: 17890-56-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...