(1R,2S)-1-Amino-2-indanol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (1R,2S)-(+)-cis-1-Amino-2-indanol, (1R,2S)-(+)-cis-1-Aminoindan-2-ol

OR2317-5G
APOSOR2317-5GEA 21.9 EUR
APOSOR2317-5G
(1R,2S)-1-Amino-2-indanol 98%
(1R,2S)-1-Amino-2,3-dihydro-1H-inden-2-ol
Formel: C₉H₁₁NO
Molecular Weight: 149,19 g/mol
Schmelzpunkt: 118…121 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00216656
CAS-Nummer: 136030-00-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR