Benzo[b]thiophen-2-ylmethanamine
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-(Aminomethyl)benzo[b]thiophene, (1-Benzothien-2-ylmethyl)amine, (Benzo[b]thiophen-2-yl)methylamine, (1-Benzothiophen-2-yl)methylamine
OR23170-250MG
APOSOR23170-250MGEA
42.2
EUR
APOSOR23170-250MG
Benzo[b]thiophen-2-ylmethanamine
Benzo[b]thiophen-2-ylmethanamine
Formel:
C₉H₉NS Molecular Weight: 163,23 g/mol Schmelzpunkt: 58…59 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00965305 CAS-Nummer: 6314-43-8 UN: 3259 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...