3-Benzo[b]thiophencarbonsäure
Lieferant: Apollo Scientific
|
Achtung
|
OR23171-1G
OR23171-250MG
OR23171-5G
APOSOR23171-1GEA
98.6
EUR
APOSOR23171-1G
APOSOR23171-250MG
APOSOR23171-5G
3-Benzo[b]thiophencarbonsäure
3-Benzo[b]thiophencarbonsäure
Formel:
C₉H₆O₂S Schmelzpunkt: 174…176 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01846406 CAS-Nummer: 5381-25-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...