3-Benzo[b]thiophenboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR23178-25G
OR23178-5G
OR23178-1G
APOSOR23178-25GEA
157
EUR
APOSOR23178-25G
APOSOR23178-5G
APOSOR23178-1G
3-Benzo[b]thiophenboronsäure 98%
3-Benzo[b]thiophenboronsäure
Formel:
C₈H₇BO₂S Schmelzpunkt: 225…230 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01114669 CAS-Nummer: 113893-08-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...