3-(Aminomethyl)benzo[b]thiophene hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(Aminomethyl)-1-benzothiophene hydrochloride, (Benzo[b]thiophen-3-yl)methylamine hydrochloride, Benzo[b]thiophen-3-ylmethanamine hydrochloride

OR23179-1G
APOSOR23179-1GEA 342 EUR
APOSOR23179-1G
3-(Aminomethyl)benzo[b]thiophene hydrochloride
3-(Aminomethyl)benzo[b]thiophene hydrochloride
Formel: C₉H₉NS·ClH
Molecular Weight: 199,7 g/mol
Schmelzpunkt: 241…244 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01566652
CAS-Nummer: 55810-74-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR