Benzo[b]thiophene-3-carboxamidine hydrochloride hydrate

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Benzothiophene-3-carboximidamide hydrochloride hydrate, 3-Carbamimidoylbenzo[b]thiophene hydrochloride hydrate

OR23181-250MG
APOSOR23181-250MGEA 92.8 EUR
APOSOR23181-250MG
Benzo[b]thiophene-3-carboxamidine hydrochloride hydrate
Benzo[b]thiophene-3-carboxamidine hydrochloride hydrate
Formel: C₉H₈N₂S·ClH·H₂O
Molecular Weight: 230,714 g/mol
Schmelzpunkt: 230 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03659730
CAS-Nummer: 465515-36-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR