4-Benzyl-2-morpholinecarboxylic acid hydrochloride

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-Benzylmorpholine-2-carboxylic acid hydrochloride

OR23182-1G OR23182-250MG OR23182-5G
APOSOR23182-1GEA 29 EUR
APOSOR23182-1G APOSOR23182-250MG APOSOR23182-5G
4-Benzyl-2-morpholinecarboxylic acid hydrochloride
4-Benzyl-2-morpholinecarboxylic acid hydrochloride
Formel: C₁₂H₁₆ClNO₃
Molecular Weight: 257.71 g/mol
Schmelzpunkt: 244…248 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02682031
CAS-Nummer: 135072-15-0
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR