4-Benzylmorpholine-2-carbonylchloride hydrochloride

Lieferant: Apollo Scientific
Achtung

OR23183-1G OR23183-100MG OR23183-250MG OR23183-500MG
APOSOR23183-1GEA 589 EUR
APOSOR23183-1G APOSOR23183-100MG APOSOR23183-250MG APOSOR23183-500MG
4-Benzylmorpholine-2-carbonylchloride hydrochloride
4-Benzylmorpholine-2-carbonylchloride hydrochloride
Formel: C₁₂H₁₅Cl₂NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03407323
CAS-Nummer: 135072-14-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR