(4-Benzyl-1,4-oxazinan-2-yl)methylamine 95+%

Lieferant: Apollo Scientific
Gefahr

Synonyme: (4-Benzylmorpholin-2-yl)methylamine

OR23186-1G
APOSOR23186-1GEA 0 EUR
APOSOR23186-1G
(4-Benzyl-1,4-oxazinan-2-yl)methylamine 95+%
(4-Benzyl-1,4-oxazinan-2-yl)methylamine
Formel: C₁₂H₁₈N₂O
Molecular Weight: 206,29 g/mol
Siedepunkt: 120…122 °C/0,5mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD02682010
CAS-Nummer: 110859-47-7
UN: 2735
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR