1,3-Dimethyl-5-(hydroxymethyl)-1H-pyrazole

Lieferant: Apollo Scientific
Achtung

Synonyme: (1,3-Dimethyl-1H-pyrazol-5-yl)methanol

OR23188-250MG
APOSOR23188-250MGEA 26.1 EUR
APOSOR23188-250MG
1,3-Dimethyl-5-(hydroxymethyl)-1H-pyrazole
1,3-Dimethyl-5-(hydroxymethyl)-1H-pyrazole
Formel: C₆H₁₀N₂O
Molecular Weight: 126,156 g/mol
Siedepunkt: 117…118 °C (0,8 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD02682037
CAS-Nummer: 57012-20-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR