5-Phenylisoxazole
Lieferant: Apollo Scientific
|
Achtung
|
OR23194-1G
OR23194-500MG
OR23194-5G
APOSOR23194-1GEA
50.9
EUR
APOSOR23194-1G
APOSOR23194-500MG
APOSOR23194-5G
5-Phenylisoxazole
5-Phenylisoxazole
Formel:
C₉H₇NO Schmelzpunkt: 23…25 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00090038 CAS-Nummer: 1006-67-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...