3-Thiophenecarbonyl chloride
Lieferant: Apollo Scientific
|
Gefahr
|
OR23199-1G
OR23199-5G
APOSOR23199-1GEA
0
EUR
APOSOR23199-1G
APOSOR23199-5G
3-Thiophenecarbonyl chloride
3-Thiophenecarbonyl chloride
Formel:
C₅H₃ClOS Schmelzpunkt: 49…51 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00130090 CAS-Nummer: 41507-35-1 EINECS: 255-420-9 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...